ENA296274877 Enamine

4-(chloromethyl)-5-methyl-2-(thiophen-3-yl)-1,3-oxazole

  • MDL number MFCD16622128
  • Cas Number : 1107663-77-3
  • Molecular Weight : 213.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65381
Availability details for
Quantity
  • Availability
  • Add to Cart