ENA296274878 Enamine

3-(2,3-dioxo-1,2,3,4-tetrahydropyrazin-1-yl)propanoic acid

  • MDL number MFCD16622129
  • Cas Number : 1240528-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65383

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65383
Availability details for
Quantity
  • Availability
  • Add to Cart