ENA296274889 Enamine

5-bromo-2-(1-chloroethyl)-1H-1,3-benzodiazole hydrochloride

  • MDL number MFCD16622145
  • Cas Number : 1240526-43-5
  • Molecular Weight : 295.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-65525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65525
Availability details for
Quantity
  • Availability
  • Add to Cart