ENA296274891 Enamine

4-amino-2-(1,3-benzothiazol-2-yl)-1,1,1-trifluorobutan-2-ol

  • MDL number MFCD16622147
  • Cas Number : 1240529-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65532
Availability details for
Quantity
  • Availability
  • Add to Cart