ENA296274896 Enamine

4-(1,3-thiazol-2-yl)piperidine dihydrochloride

  • MDL number MFCD16622152
  • Cas Number : 1240529-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65568

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65568
Availability details for
Quantity
  • Availability
  • Add to Cart