ENA296274899 Enamine

1-(1-aminopentan-3-yl)pyrrolidin-2-one hydrochloride

  • MDL number MFCD16622155
  • Cas Number : 1240529-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65580
Availability details for
Quantity
  • Availability
  • Add to Cart