ENA296274901 Enamine

potassium 5-hydroxypentanoate

  • MDL number MFCD16622157
  • Cas Number : 1240528-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65598
Availability details for
Quantity
  • Availability
  • Add to Cart