ENA296274902 Enamine

4-chloro-6-cyclobutylpyrimidine

  • MDL number MFCD16622158
  • Cas Number : 768397-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65613
Availability details for
Quantity
  • Availability
  • Add to Cart