ENA296274903 Enamine

N-(5-bromopyridin-2-yl)-2-(methylamino)acetamide hydrochloride

  • MDL number MFCD16622159
  • Cas Number : 1240528-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 272...274
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65619

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65619
Availability details for
Quantity
  • Availability
  • Add to Cart