ENA296274909 Enamine

4-amino-N-phenylbutanamide hydrochloride

  • MDL number MFCD16622166
  • Cas Number : 115022-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65638
Availability details for
Quantity
  • Availability
  • Add to Cart