ENA296274912 Enamine

3-(ethylamino)-2-methylpropanoic acid hydrochloride

  • MDL number MFCD16622169
  • Cas Number : 1240529-23-0
  • Molecular Weight : 167.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65645
Availability details for
Quantity
  • Availability
  • Add to Cart