ENA296274914 Enamine

1-(4-aminopiperidin-1-yl)-3,3,3-trifluoropropan-1-one hydrochloride

  • MDL number MFCD16622171
  • Cas Number : 1187160-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65691
Availability details for
Quantity
  • Availability
  • Add to Cart