ENA296274915 Enamine

3-amino-1,1-difluorobutan-2-ol hydrochloride

  • MDL number MFCD16622172
  • Cas Number : 1240528-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65696

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65696
Availability details for
Quantity
  • Availability
  • Add to Cart