ENA296274920 Enamine

N-(2-aminophenyl)-decahydroquinoline-2-carboxamide

  • MDL number MFCD16622180
  • Cas Number : 1251923-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65710
Availability details for
Quantity
  • Availability
  • Add to Cart