ENA296274923 Enamine

2-chloroethyl N-(pyridin-3-yl)carbamate hydrochloride

  • MDL number MFCD16622183
  • Cas Number : 1240528-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65751
Availability details for
Quantity
  • Availability
  • Add to Cart