ENA296274926 Enamine

5-(2-methyl-1,3-thiazol-4-yl)furan-3-carboxylic acid

  • MDL number MFCD16622187
  • Cas Number : 1240526-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65785

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65785
Availability details for
Quantity
  • Availability
  • Add to Cart