ENA296274936 Enamine

(2,5-difluorophenyl)(1H-1,2,3,4-tetrazol-5-yl)methanamine hydrochloride

  • MDL number MFCD16622197
  • Cas Number : 1240528-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65859
Availability details for
Quantity
  • Availability
  • Add to Cart