ENA296274943 Enamine

1-{4-[(4-methylphenyl)methoxy]phenyl}propan-2-amine hydrochloride

  • MDL number MFCD16622206
  • Cas Number : 1240528-44-2
  • Molecular Weight : 291.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65990
Availability details for
Quantity
  • Availability
  • Add to Cart