ENA296274946 Enamine

3-(3-aminophenyl)-4,5-dihydro-1,2,4-oxadiazol-5-one hydrochloride

  • MDL number MFCD16622209
  • Cas Number : 145878-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66033
Availability details for
Quantity
  • Availability
  • Add to Cart