ENA296274949 Enamine

4-bromo-3-iodoaniline

  • MDL number MFCD16622212
  • Cas Number : 63037-64-9
  • Molecular Weight : 297.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-77,5
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66174

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P310,P302+P352,P304+P340,P312,P330,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-66174
pictograms GHS06
hstatements H301,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart