ENA296274967 Enamine

2-[5-(piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyridine

  • MDL number MFCD10034869
  • Cas Number : 1018593-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64826

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64826
Availability details for
Quantity
  • Availability
  • Add to Cart