ENA296274968 Enamine

tert-butyl N-[1-(N'-hydroxycarbamimidoyl)cyclopentyl]carbamate

  • Cas Number : 1240562-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53826
Availability details for
Quantity
  • Availability
  • Add to Cart