ENA296274970 Enamine

N-(3-aminopropyl)-4-bromobenzamide

  • MDL number MFCD01069850
  • Cas Number : 1016790-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64836
Availability details for
Quantity
  • Availability
  • Add to Cart