ENA296274971 Enamine

3-azaspiro[5.5]undecane

  • MDL number MFCD00011572
  • Cas Number : 180-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64837

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-64837
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart