ENA296274974 Enamine

[1-(5-chloro-2-methylphenyl)pyrrolidin-3-yl]methanamine

  • MDL number MFCD09934776
  • Cas Number : 1017428-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64858
Availability details for
Quantity
  • Availability
  • Add to Cart