ENA296274979 Enamine

4-amino-N,2-dimethylbenzene-1-sulfonamide

  • MDL number MFCD11621467
  • Cas Number : 101252-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64872
Availability details for
Quantity
  • Availability
  • Add to Cart