ENA296274980 Enamine

4-[(2-methoxyphenyl)methyl]piperidine hydrochloride

  • MDL number MFCD08437508
  • Cas Number : 37581-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64876
Availability details for
Quantity
  • Availability
  • Add to Cart