ENA296274983 Enamine

5-phenylazepan-2-one

  • MDL number MFCD12174726
  • Cas Number : 7500-39-2
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-54450

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54450
Availability details for
Quantity
  • Availability
  • Add to Cart