ENA296274987 Enamine

1-[(2-aminophenyl)methyl]pyrrolidine-2-carboxamide

  • MDL number MFCD11173578
  • Cas Number : 1251922-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64904
Availability details for
Quantity
  • Availability
  • Add to Cart