ENA296274988 Enamine

4-bromo-3-(propan-2-yl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD13704082
  • Cas Number : 1257878-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195...197
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54548

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54548
Availability details for
Quantity
  • Availability
  • Add to Cart