ENA296274991 Enamine

2-(phenylamino)butan-1-ol

  • MDL number MFCD12781332
  • Cas Number : 51170-06-0
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64909

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64909
Availability details for
Quantity
  • Availability
  • Add to Cart