ENA296275000 Enamine

1-(oxolan-2-yl)propan-2-amine

  • MDL number MFCD12154009
  • Cas Number : 100868-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64935

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64935
Availability details for
Quantity
  • Availability
  • Add to Cart