ENA296275004 Enamine

N-(1,2,3-thiadiazol-4-ylmethyl)pyridin-3-amine

  • MDL number MFCD12112953
  • Cas Number : 1156894-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64944

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64944
Availability details for
Quantity
  • Availability
  • Add to Cart