ENA296275009 Enamine

3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanethioamide

  • MDL number MFCD02932980
  • Cas Number : 57001-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186-188
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55131
Availability details for
Quantity
  • Availability
  • Add to Cart