ENA296275011 Enamine

1-(bromomethyl)-4-(propan-2-yloxy)benzene

  • MDL number MFCD11180378
  • Cas Number : 72729-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Dry Ice
Product Link https://www.enaminestore.com/catalog/EN300-64984

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-64984
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart