ENA296275014 Enamine

3-(4-methyl-1,3-thiazol-2-yl)propan-1-amine

  • MDL number MFCD09891572
  • Cas Number : 112086-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64988

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64988
Availability details for
Quantity
  • Availability
  • Add to Cart