ENA296275017 Enamine

4-methoxy-3-(5-methyl-1H-1,2,3,4-tetrazol-1-yl)aniline

  • MDL number MFCD12091841
  • Cas Number : 857751-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64992

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64992
Availability details for
Quantity
  • Availability
  • Add to Cart