ENA296275019 Enamine

3-[(1H-indazol-7-yl)sulfamoyl]thiophene-2-carboxylic acid

  • MDL number MFCD13691618
  • Cas Number : 1154239-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64994
Availability details for
Quantity
  • Availability
  • Add to Cart