ENA296275020 Enamine

3-amino-1,1,1-trifluoro-2-phenylpropan-2-ol hydrochloride

  • MDL number MFCD16617240
  • Cas Number : 1240527-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218...220
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64995

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64995
Availability details for
Quantity
  • Availability
  • Add to Cart