ENA296275021 Enamine

2-amino-N-[1-(4-cyanophenyl)ethyl]-5-fluorobenzamide

  • MDL number MFCD13773986
  • Cas Number : 1154287-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64996

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64996
Availability details for
Quantity
  • Availability
  • Add to Cart