ENA296275025 Enamine

2-(3-fluorophenyl)-2-(methylamino)acetamide

  • MDL number MFCD12151213
  • Cas Number : 1218316-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65001
Availability details for
Quantity
  • Availability
  • Add to Cart