ENA296275027 Enamine

3-(4-methyl-1,3-thiazol-2-yl)aniline

  • MDL number MFCD06739764
  • Cas Number : 134811-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82...84
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65008

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-65008
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart