ENA296275044 Enamine

4-(3-phenylpropyl)aniline

  • MDL number MFCD11976349
  • Cas Number : 80861-22-9
  • Molecular Weight : 211.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-65064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65064
Availability details for
Quantity
  • Availability
  • Add to Cart