ENA296275045 Enamine

1-(3-bromophenyl)-4,4-dimethylpentan-3-one

  • MDL number MFCD11655204
  • Cas Number : 1099653-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65065

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65065
Availability details for
Quantity
  • Availability
  • Add to Cart