ENA296275048 Enamine

N-(1-methylpiperidin-4-yl)-1H-indazol-6-amine

  • MDL number MFCD01308752
  • Cas Number : 416880-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65070
Availability details for
Quantity
  • Availability
  • Add to Cart