ENA296275051 Enamine

1,3-dimethyl-5-(1H-1,2,4-triazol-1-yl)-1H-pyrazol-4-amine

  • MDL number MFCD12136910
  • Cas Number : 1157096-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96...98
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65073

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65073
Availability details for
Quantity
  • Availability
  • Add to Cart