ENA296275056 Enamine

5-(2,2,2-trifluoroethyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09814806
  • Cas Number : 1016735-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65078

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65078
Availability details for
Quantity
  • Availability
  • Add to Cart