ENA296275057 Enamine

3-(4-amino-2-methoxyphenyl)-1,3-oxazolidin-2-one

  • MDL number MFCD11167811
  • Cas Number : 31847-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55723

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55723
Availability details for
Quantity
  • Availability
  • Add to Cart