ENA296275058 Enamine

3-(1H-1,2,3-benzotriazole-5-sulfonamido)benzoic acid

  • MDL number MFCD05262721
  • Cas Number : 743440-18-8
  • Molecular Weight : 318.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-65081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65081
Availability details for
Quantity
  • Availability
  • Add to Cart