ENA296275059 Enamine

6-cyclopropyl-1-[(5-methylfuran-2-yl)methyl]-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09731387
  • Cas Number : 953905-94-7
  • Molecular Weight : 297.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55917
Availability details for
Quantity
  • Availability
  • Add to Cart